siRNA Details
Gene Symbol: ZBTB39 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014830.2 | NP_055645.1 | 2 | 3368 | ||
GenBank mRNA | AB002350.1 | - | 2 | 3301 | ||
BC140890.1 | - | 1 | 3271 | 9021758 | ||
BC146867.1 | - | 1 | 3271 | 9056795 | ||
BC152461.1 | - | 2 | 3301 | 8862597 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
