siRNA Details
Gene Symbol: ZBTB39 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014830.2 | NP_055645.1 | 2 | 4898 | ||
GenBank mRNA | AB002350.1 | - | 2 | 4831 | ||
BC152461.1 | - | 2 | 4831 | 8862597 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
