siRNA Details
Gene Symbol: COL6A2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001849.3 | NP_001840.3 | 25 | 1997 | ||
NM_058174.2 | NP_478054.2 | 25 | 1997 | |||
NM_058175.2 | NP_478055.2 | 25 | 1997 | |||
XM_011529451.1 | XP_011527753.1 | 25 | 2288 | |||
GenBank mRNA | AY029208.1 | - | 24 | 1936 | ||
BC002484.2 | - | 5 | 314 | |||
BC065509.1 | - | 25 | 1983 | 5723962 | ||
BT006913.1 | - | 2 | 133 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
