siRNA Details
Gene Symbol: CARMIL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013838.1 | NP_001013860.1 | 29 | 3148 | ||
NM_001317026.1 | NP_001303955.1 | 30 | 3040 | |||
XM_011522874.1 | XP_011521176.1 | 30 | 3178 | |||
XM_011522875.2 | XP_011521177.2 | 30 | 3178 | |||
XM_017022953.1 | XP_016878442.1 | 30 | 3178 | |||
GenBank mRNA | AB113647.1 | - | 29 | 3148 | ||
AK090421.1 | - | 5 | 4451 | |||
AL832446.1 | - | 28 | 2979 | |||
BC043154.1 | - | 1 | 106 | |||
FJ026014.1 | - | 30 | 2920 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
