siRNA Details
Gene Symbol: ZNF90 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007138.1 | NP_009069.1 | 4 | 435 | ||
GenBank mRNA | BC137211.1 | - | 4 | 362 | 9021208 | |
CN386341.1 | - | 4 | 362 | |||
M61870.1 | - | 4 | 417 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
