siRNA Details
Gene Symbol: TTYH1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005367.2 | NP_001005367.1 | 13 | 1799 | ||
NM_001201461.1 | NP_001188390.1 | 14 | 1848 | |||
NM_020659.3 | NP_065710.1 | 14 | 1845 | |||
XM_017027025.1 | XP_016882514.1 | 13 | 1643 | |||
XM_017027026.1 | XP_016882515.1 | 14 | 1689 | |||
GenBank mRNA | AB162930.1 | - | 13 | 1681 | ||
AB188496.1 | - | 14 | 1848 | |||
AF177909.1 | - | 14 | 1758 | |||
BC011347.1 | - | 13 | 1704 | 4215136 | ||
BC019358.1 | - | 11 | 1181 | 3960435 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
