siRNA Details
Gene Symbol: LILRA5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021250.3 | NP_067073.1 | 4 | 350 | ||
NM_181879.2 | NP_870994.1 | 4 | 350 | |||
NM_181985.3 | NP_871714.1 | 3 | 314 | |||
NM_181986.2 | NP_871715.1 | 3 | 314 | |||
GenBank mRNA | AF212842.1 | - | 1 | 64 | ||
AF324830.1 | - | 4 | 350 | |||
AF499916.1 | - | 4 | 293 | |||
AF499917.1 | - | 3 | 249 | |||
AF499918.1 | - | 4 | 309 | |||
AF499919.1 | - | 3 | 266 | |||
EU915611.1 | - | 4 | 290 | |||
EU915612.1 | - | 3 | 254 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
