siRNA Details
Gene Symbol: LILRA5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021250.3 | NP_067073.1 | 7 | 1049 | ||
NM_181985.3 | NP_871714.1 | 6 | 1013 | |||
GenBank mRNA | AF212842.1 | - | 4 | 763 | ||
AF324830.1 | - | 7 | 1049 | |||
AF499916.1 | - | 7 | 992 | |||
AF499917.1 | - | 6 | 948 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
