siRNA Details
Gene Symbol: HDGFRP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001520.2 | NP_001001520.1 | 13, 14 | 1734 | ||
NM_032631.3 | NP_116020.1 | 13, 14 | 1734 | |||
XM_011528375.2 | XP_011526677.1 | 13, 14 | 2094 | |||
XM_011528376.2 | XP_011526678.1 | 13, 14 | 2091 | |||
XM_011528377.2 | XP_011526679.1 | 13, 14 | 2094 | |||
GenBank mRNA | AB208892.1 | - | 11, 12 | 2472 | ||
AF294267.1 | - | 13, 14 | 1641 | |||
AK027353.1 | - | 1, 2 | 121 | |||
AY358600.1 | - | 13, 14 | 1643 | |||
BC000755.2 | - | 13, 14 | 1624 | 3507276 | ||
BC009449.1 | - | 10, 11 | 1273 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
