siRNA Details
Gene Symbol: ANGPTL4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039667.2 | NP_001034756.1 | 6 | 1764 | ||
NM_139314.2 | NP_647475.1 | 7 | 1878 | |||
NR_104213.1 | 3 | 780 | ||||
XM_005272484.3 | XP_005272541.1 | 7 | 1930 | |||
XM_005272485.3 | XP_005272542.1 | 6 | 1816 | |||
GenBank mRNA | AB056477.1 | - | 7 | 1852 | ||
AF153606.1 | - | 8 | 1773 | |||
AF202636.1 | - | 7 | 1854 | |||
AK222489.1 | - | 7 | 1850 | |||
AK222524.1 | - | 7 | 1848 | |||
AW954426.1 | - | 1 | 569 | |||
AY358275.1 | - | 7 | 1846 | |||
BC023647.2 | - | 7 | 1822 | 5088323 | ||
BP397348.1 | - | 1 | 306 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
