siRNA Details
Gene Symbol: UQCRFS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK291134.1 | - | 1 | 61 | ||
BC000649.1 | - | 1 | 28 | 3345521 | ||
BC010035.2 | - | 1 | 43 | 3538225 | ||
BC067832.1 | - | 1 | 42 | 5285759 | ||
BF979273.1 | - | 1 | 143 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
