siRNA Details
Gene Symbol: SMAD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001003688.1 | NP_001003688.1 | 6 | 1366 | ||
NM_005900.2 | NP_005891.1 | 6 | 1542 | |||
XM_005262992.3 | XP_005263049.1 | 6 | 1335 | |||
XM_006714217.2 | XP_006714280.1 | 6 | 1397 | |||
XM_011531961.1 | XP_011530263.1 | 6 | 1357 | |||
XM_011531962.1 | XP_011530264.1 | 6 | 1402 | |||
XM_011531963.1 | XP_011530265.1 | 6 | 1524 | |||
XM_011531964.1 | XP_011530266.1 | 6 | 1554 | |||
GenBank mRNA | AK096624.1 | - | 2 | 2877 | ||
AK293055.1 | - | 6 | 1668 | |||
BC001878.1 | - | 6 | 1467 | 3546771 | ||
BT007386.1 | - | 5 | 1126 | |||
U54826.1 | - | 6 | 1366 | |||
U57456.1 | - | 5 | 1278 | |||
U59423.1 | - | 6 | 1558 | |||
U59912.1 | - | 6 | 1367 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
