siRNA Details
Gene Symbol: UMOD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008389.2 | NP_001008390.1 | 8 | 1909 | ||
NM_001278614.1 | NP_001265543.1 | 9 | 2071 | |||
NM_003361.3 | NP_003352.2 | 8 | 1972 | |||
XM_011545934.1 | XP_011544236.1 | 9 | 2050 | |||
XM_011545935.1 | XP_011544237.1 | 10 | 2109 | |||
XM_011545936.1 | XP_011544238.1 | 10 | 2046 | |||
XM_011545937.1 | XP_011544239.1 | 10 | 1936 | |||
XM_011545938.1 | XP_011544240.1 | 9 | 1968 | |||
XM_011545940.1 | XP_011544242.1 | 9 | 1986 | |||
XM_017023641.1 | XP_016879130.1 | 8 | 1909 | |||
GenBank mRNA | AK055722.1 | - | 9 | 1675 | ||
AK091961.1 | - | 9 | 1903 | |||
AK096043.1 | - | 9 | 1552 | |||
AK127643.1 | - | 9 | 1405 | |||
AK127648.1 | - | 9 | 1668 | |||
BC035975.1 | - | 9 | 1658 | 4556427 | ||
M15881.1 | - | 8 | 1848 | |||
M17778.1 | - | 7 | 1785 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
