siRNA Details
Gene Symbol: UMOD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008389.2 | NP_001008390.1 | 9 | 2002 | ||
NM_001278614.1 | NP_001265543.1 | 10 | 2164 | |||
NM_003361.3 | NP_003352.2 | 9 | 2065 | |||
XM_011545934.1 | XP_011544236.1 | 10 | 2143 | |||
XM_011545935.1 | XP_011544237.1 | 11 | 2202 | |||
XM_011545936.1 | XP_011544238.1 | 11 | 2139 | |||
XM_011545937.1 | XP_011544239.1 | 11 | 2029 | |||
XM_011545938.1 | XP_011544240.1 | 10 | 2061 | |||
XM_011545940.1 | XP_011544242.1 | 10 | 2079 | |||
XM_017023641.1 | XP_016879130.1 | 9 | 2002 | |||
GenBank mRNA | AK055722.1 | - | 10 | 1768 | ||
AK091961.1 | - | 10 | 1996 | |||
AK096043.1 | - | 10 | 1645 | |||
AK127643.1 | - | 10 | 1498 | |||
AK127648.1 | - | 10 | 1761 | |||
BC035975.1 | - | 10 | 1751 | 4556427 | ||
M15881.1 | - | 9 | 1941 | |||
M17778.1 | - | 8 | 1878 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
