siRNA Details
Gene Symbol: OR5B2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005566.2 | NP_001005566.1 | 1 | 206 | ||
GenBank mRNA | BC136922.1 | - | 1 | 209 | 9020914 | |
BC136923.1 | - | 1 | 203 | 9020915 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
