siRNA Details
Gene Symbol: SGCD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000337.5 | NP_000328.2 | 9 | 5366 | ||
NM_001128209.1 | NP_001121681.1 | 8 | 5320 | |||
GenBank mRNA | BX537948.1 | - | 8 | 5332 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
