siRNA Details
Gene Symbol: ARL13B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001174150.1 | NP_001167621.1 | 4, 5 | 782 | ||
NM_001174151.1 | NP_001167622.1 | 3, 4 | 711 | |||
NM_001321328.1 | NP_001308257.1 | 5, 6 | 904 | |||
NM_144996.3 | NP_659433.2 | 2, 3 | 461 | |||
NM_182896.2 | NP_878899.1 | 4, 5 | 782 | |||
NR_033427.1 | 3, 4 | 537 | ||||
NR_135621.1 | 3, 4 | 528 | ||||
XM_006713532.3 | XP_006713595.1 | 5, 6 | 901 | |||
XM_011512532.2 | XP_011510834.1 | 5, 6 | 810 | |||
XM_011512533.2 | XP_011510835.1 | 6, 7 | 618 | |||
XM_011512534.2 | XP_011510836.1 | 6, 7 | 936 | |||
XM_011512535.2 | XP_011510837.1 | 5, 6 | 795 | |||
XM_017005853.1 | XP_016861342.1 | 5, 6 | 812 | |||
GenBank mRNA | AK296994.1 | - | 4, 5 | 760 | ||
AK299284.1 | - | 3, 4 | 521 | |||
AL713789.1 | - | 3, 4 | 653 | |||
BC015439.1 | - | 2, 3 | 421 | |||
BC094725.1 | - | 2, 3 | 456 | 4818884 | ||
BC104035.1 | - | 4, 5 | 772 | 40026398 | ||
BC104036.1 | - | 4, 5 | 772 | 40026399 | ||
BG705058.1 | - | 2, 3 | 466 | |||
CR936753.1 | - | 2, 3 | 459 | |||
CR936756.1 | - | 3, 4 | 416 | |||
CR936763.1 | - | 5, 6 | 856 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
