siRNA Details
Gene Symbol: ARL13B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001174150.1 | NP_001167621.1 | 10 | 1565 | ||
NM_001174151.1 | NP_001167622.1 | 9 | 1494 | |||
NM_001321328.1 | NP_001308257.1 | 11 | 1687 | |||
NM_144996.3 | NP_659433.2 | 8 | 1244 | |||
NM_182896.2 | NP_878899.1 | 10 | 1565 | |||
NR_033427.1 | 9 | 1320 | ||||
NR_135621.1 | 9 | 1311 | ||||
XM_006713532.3 | XP_006713595.1 | 11 | 1684 | |||
XM_011512532.2 | XP_011510834.1 | 11 | 1593 | |||
XM_011512533.2 | XP_011510835.1 | 12 | 1401 | |||
XM_011512534.2 | XP_011510836.1 | 12 | 1719 | |||
XM_011512535.2 | XP_011510837.1 | 11 | 1578 | |||
XM_017005853.1 | XP_016861342.1 | 11 | 1595 | |||
GenBank mRNA | AK296994.1 | - | 10 | 1543 | ||
AK299284.1 | - | 9 | 1304 | |||
AL713789.1 | - | 9 | 1437 | |||
BC094725.1 | - | 8 | 1239 | 4818884 | ||
BC104035.1 | - | 10 | 1555 | 40026398 | ||
BC104036.1 | - | 10 | 1555 | 40026399 | ||
CR936753.1 | - | 8 | 1242 | |||
CR936756.1 | - | 9 | 1199 | |||
CR936763.1 | - | 11 | 1639 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
