siRNA Details
Gene Symbol: ARL13B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001174150.1 | NP_001167621.1 | 4 | 708 | ||
NM_001174151.1 | NP_001167622.1 | 3 | 637 | |||
NM_001321328.1 | NP_001308257.1 | 5 | 830 | |||
NM_144996.3 | NP_659433.2 | 2 | 387 | |||
NM_182896.2 | NP_878899.1 | 4 | 708 | |||
NR_033427.1 | 3 | 458 | ||||
NR_135621.1 | 3 | 454 | ||||
XM_006713532.3 | XP_006713595.1 | 5 | 827 | |||
XM_011512532.2 | XP_011510834.1 | 5 | 736 | |||
XM_011512533.2 | XP_011510835.1 | 6 | 544 | |||
XM_011512534.2 | XP_011510836.1 | 6 | 862 | |||
XM_011512535.2 | XP_011510837.1 | 5 | 721 | |||
XM_017005853.1 | XP_016861342.1 | 5 | 738 | |||
GenBank mRNA | AK296994.1 | - | 4 | 686 | ||
AK299284.1 | - | 3 | 442 | |||
AL713789.1 | - | 3 | 579 | |||
BC015439.1 | - | 2 | 347 | |||
BC094725.1 | - | 2 | 382 | 4818884 | ||
BC104035.1 | - | 4 | 698 | 40026398 | ||
BC104036.1 | - | 4 | 698 | 40026399 | ||
CR936753.1 | - | 2 | 385 | |||
CR936756.1 | - | 3 | 342 | |||
CR936763.1 | - | 5 | 782 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
