siRNA Details
Gene Symbol: CNR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001160226.1 | NP_001153698.1 | 3 | 1759 | ||
NM_001160258.1 | NP_001153730.1 | 4 | 1797 | |||
NM_001160259.1 | NP_001153731.1 | 2 | 1672 | |||
NM_016083.4 | NP_057167.2 | 2 | 1616 | |||
NM_033181.3 | NP_149421.2 | 2 | 1271 | |||
XM_006715330.3 | XP_006715393.1 | 2 | 1563 | |||
XM_011535424.1 | XP_011533726.1 | 3 | 2103 | |||
XM_011535425.1 | XP_011533727.1 | 4 | 2141 | |||
XM_011535426.1 | XP_011533728.1 | 5 | 2299 | |||
XM_011535428.1 | XP_011533730.1 | 3 | 1950 | |||
XM_017010238.1 | XP_016865727.1 | 4 | 4927 | |||
XM_017010239.1 | XP_016865728.1 | 3 | 3250 | |||
XM_017010240.1 | XP_016865729.1 | 2 | 1610 | |||
GenBank mRNA | AF107262.1 | - | 1 | 1316 | ||
AK313908.1 | - | 2 | 1437 | |||
AY766182.1 | - | 2 | 1208 | |||
AY995204.1 | - | 1 | 1307 | |||
BC074811.2 | - | 1 | 1345 | 30915316 | ||
BC074812.2 | - | 1 | 1345 | 30915532 | ||
BC095513.1 | - | 1 | 1304 | |||
BC100968.1 | - | 1 | 1307 | 40004970 | ||
BC100969.1 | - | 1 | 1307 | 40004975 | ||
BC100970.1 | - | 1 | 1307 | 40004978 | ||
BC100971.1 | - | 1 | 1307 | 40004979 | ||
DQ067455.1 | - | 1 | 1307 | |||
KC292264.1 | - | 2 | 1271 | |||
X54937.1 | - | 2 | 1455 | |||
X81120.1 | - | 1 | 1398 | |||
X81121.1 | - | 2 | 1140 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Set Membership: |
![]() |
Gene Ontology Categories:
