siRNA Details
Gene Symbol: CNR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001160226.1 | NP_001153698.1 | 3 | 465 | ||
NM_001160258.1 | NP_001153730.1 | 4 | 503 | |||
NM_001160259.1 | NP_001153731.1 | 2 | 378 | |||
NM_016083.4 | NP_057167.2 | 2 | 322 | |||
NM_033181.3 | NP_149421.2 | 1 | 76 | |||
XM_006715330.3 | XP_006715393.1 | 2 | 269 | |||
XM_011535424.1 | XP_011533726.1 | 3 | 809 | |||
XM_011535425.1 | XP_011533727.1 | 4 | 847 | |||
XM_011535426.1 | XP_011533728.1 | 5 | 1005 | |||
XM_011535428.1 | XP_011533730.1 | 3 | 656 | |||
XM_017010238.1 | XP_016865727.1 | 4 | 3633 | |||
XM_017010239.1 | XP_016865728.1 | 3 | 1956 | |||
XM_017010240.1 | XP_016865729.1 | 2 | 316 | |||
GenBank mRNA | AF107262.1 | - | 1 | 22 | ||
AK313908.1 | - | 2 | 143 | |||
AY766182.1 | - | 1 | 13 | |||
AY995204.1 | - | 1 | 13 | |||
BC074811.2 | - | 1 | 51 | 30915316 | ||
BC074812.2 | - | 1 | 51 | 30915532 | ||
BC100968.1 | - | 1 | 13 | 40004970 | ||
BC100969.1 | - | 1 | 13 | 40004975 | ||
BC100970.1 | - | 1 | 13 | 40004978 | ||
BC100971.1 | - | 1 | 13 | 40004979 | ||
DQ067455.1 | - | 1 | 13 | |||
KC292264.1 | - | 1 | 76 | |||
X54937.1 | - | 2 | 161 | |||
X81120.1 | - | 1 | 104 | |||
X81121.1 | - | 1 | 13 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Set Membership: |
![]() |
Gene Ontology Categories:
