siRNA Details
Gene Symbol: SYCE1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001143763.1 | NP_001137235.1 | 6 | 464 | ||
NM_001143764.1 | NP_001137236.1 | 6 | 464 | |||
NM_130784.2 | NP_570140.1 | 6 | 387 | |||
GenBank mRNA | AK314978.1 | - | 6 | 414 | ||
AY027807.1 | - | 6 | 387 | |||
AY027808.1 | - | 3 | 1323 | |||
BC034821.1 | - | 6 | 414 | 5164386 | ||
DB053356.1 | - | 6 | 464 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
