siRNA Details
Gene Symbol: PHF14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014660.3 | NP_055475.2 | 4 | 1402 | ||
NR_033435.1 | 3 | 614 | ||||
NR_033436.1 | 3 | 614 | ||||
XM_017012811.1 | XP_016868300.1 | 4 | 1440 | |||
XM_017012812.1 | XP_016868301.1 | 4 | 1440 | |||
XM_017012813.1 | XP_016868302.1 | 4 | 1439 | |||
XM_017012814.1 | XP_016868303.1 | 4 | 1439 | |||
XM_017012815.1 | XP_016868304.1 | 4 | 1439 | |||
XM_017012816.1 | XP_016868305.1 | 4 | 1438 | |||
XM_017012817.1 | XP_016868306.1 | 3 | 649 | |||
GenBank mRNA | AB018326.1 | - | 4 | 1357 | ||
AK294419.1 | - | 3 | 614 | |||
AK295461.1 | - | 3 | 597 | |||
BC152414.1 | - | 4 | 1357 | 8862519 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
