siRNA Details
Gene Symbol: HIST1H2AE | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021052.2 | NP_066390.1 | 1 | 110 | ||
GenBank mRNA | AK311824.1 | - | 1 | 109 | ||
BC093836.1 | - | 1 | 81 | 7939681 | ||
BC093862.1 | - | 1 | 81 | 7939707 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
