siRNA Details
Gene Symbol: ZNF3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278284.1 | NP_001265213.1 | 3 | 478 | ||
NM_001278287.1 | NP_001265216.1 | 3 | 394 | |||
NM_001278290.1 | NP_001265219.1 | 2 | 239 | |||
NM_001318135.1 | NP_001305064.1 | 3 | 356 | |||
NM_017715.3 | NP_060185.2 | 3 | 360 | |||
NM_032924.4 | NP_116313.3 | 3 | 360 | |||
XM_006716111.1 | XP_006716174.1 | 3 | 340 | |||
XM_017012582.1 | XP_016868071.1 | 3 | 354 | |||
GenBank mRNA | AF193057.1 | - | 3 | 323 | ||
AF217988.1 | - | 3 | 324 | |||
AK000223.1 | - | 1 | 95 | |||
AK297625.1 | - | 3 | 322 | |||
AK309719.1 | - | 3 | 666 | |||
BC011887.2 | - | 3 | 284 | 3831842 | ||
BC013603.1 | - | 3 | 303 | 3865426 | ||
BC025265.1 | - | 3 | 293 | |||
BQ071498.1 | - | 3 | 190 | |||
BX647182.1 | - | 2 | 1001 | |||
DC347416.1 | - | 3 | 360 | |||
DC384519.1 | - | 3 | 360 | |||
HY096067.1 | - | 2 | 218 | |||
X60153.1 | - | 1 | 33 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
