siRNA Details
Gene Symbol: ZNF3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278284.1 | NP_001265213.1 | 4 | 563 | ||
NM_001278287.1 | NP_001265216.1 | 4 | 479 | |||
NM_001278290.1 | NP_001265219.1 | 3 | 324 | |||
NM_001278291.1 | NP_001265220.1 | 2 | 193 | |||
NM_001278292.1 | NP_001265221.1 | 3 | 314 | |||
NM_001318135.1 | NP_001305064.1 | 4 | 441 | |||
NM_001318136.1 | NP_001305065.1 | 2 | 193 | |||
NM_001318137.1 | NP_001305066.1 | 2 | 193 | |||
NM_017715.3 | NP_060185.2 | 4 | 445 | |||
NM_032924.4 | NP_116313.3 | 4 | 445 | |||
XM_006716111.1 | XP_006716174.1 | 4 | 499 | |||
XM_017012582.1 | XP_016868071.1 | 4 | 513 | |||
XM_017012583.1 | XP_016868072.1 | 2 | 662 | |||
GenBank mRNA | AF193057.1 | - | 4 | 408 | ||
AF217988.1 | - | 4 | 409 | |||
AK000223.1 | - | 2 | 180 | |||
AK091984.1 | - | 2 | 165 | |||
AK297625.1 | - | 4 | 407 | |||
AK309719.1 | - | 4 | 751 | |||
AK315479.1 | - | 2 | 159 | |||
BC011887.2 | - | 4 | 369 | 3831842 | ||
BC013603.1 | - | 4 | 388 | 3865426 | ||
BC025265.1 | - | 4 | 378 | |||
BQ071498.1 | - | 4 | 275 | |||
BX647182.1 | - | 3 | 1086 | |||
CB129643.1 | - | 2 | 149 | |||
CB988353.1 | - | 2 | 193 | |||
DA443888.1 | - | 2 | 179 | |||
DA623743.1 | - | 3 | 288 | |||
DC384519.1 | - | 4 | 445 | |||
X60153.1 | - | 2 | 118 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
