siRNA Details
Gene Symbol: EGLN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053046.3 | NP_444274.1 | 5, 6 | 1609 | ||
NM_080732.3 | NP_542770.2 | 5, 6 | 1519 | |||
GenBank mRNA | AJ310544.1 | - | 4, 5 | 1461 | ||
AK025396.1 | - | 1, 2 | 40 | |||
AK026863.1 | - | 2, 3 | 764 | |||
AK074408.1 | - | 5, 6 | 1448 | |||
AK098182.1 | - | 4, 5 | 2193 | |||
AL133009.1 | - | 2, 3 | 3267 | |||
AL832506.1 | - | 5, 6 | 1475 | |||
AY040565.1 | - | 4, 5 | 1453 | |||
BC001723.1 | - | 5, 6 | 1422 | |||
BC036051.1 | - | 5, 6 | 1523 | 5270945 |
Gene Symbol: RAB4B-EGLN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_037791.1 | 11, 12 | 2213 | |||
GenBank mRNA | AK291385.1 | - | 11, 12 | 2161 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
