siRNA Details
Gene Symbol: EGLN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053046.3 | NP_444274.1 | 4 | 1464 | ||
NM_080732.3 | NP_542770.2 | 4 | 1374 | |||
GenBank mRNA | AJ310544.1 | - | 3 | 1316 | ||
AK026863.1 | - | 2 | 410 | |||
AK074408.1 | - | 4 | 1303 | |||
AK098182.1 | - | 3 | 2048 | |||
AL133009.1 | - | 1 | 3122 | |||
AL832506.1 | - | 4 | 1330 | |||
AY040565.1 | - | 3 | 1308 | |||
BC001723.1 | - | 4 | 1277 | |||
BC036051.1 | - | 4 | 1378 | 5270945 |
Gene Symbol: RAB4B-EGLN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_037791.1 | 10 | 2068 | |||
GenBank mRNA | AK291385.1 | - | 10 | 2016 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
