siRNA Details
Gene Symbol: CPLX1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006651.3 | NP_006642.1 | 3, 4 | 403 | ||
XM_011513391.1 | XP_011511693.1 | 2, 3 | 334 | |||
GenBank mRNA | AB593095.1 | - | 3, 4 | 380 | ||
AF022383.1 | - | 2, 3 | 320 | |||
AK126578.1 | - | 4, 5 | 275 | |||
AK311921.1 | - | 3, 4 | 375 | |||
BC002471.2 | - | 3, 4 | 403 | 3349779 | ||
BT007029.1 | - | 2, 3 | 212 | |||
KJ534815.1 | - | 2, 3 | 212 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
