siRNA Details
Gene Symbol: CPLX1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006651.3 | NP_006642.1 | 4 | 1614 | ||
XM_011513391.1 | XP_011511693.1 | 3 | 1545 | |||
GenBank mRNA | AB593095.1 | - | 4 | 1591 | ||
AK126578.1 | - | 5 | 1486 | |||
BC002471.2 | - | 4 | 1614 | 3349779 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
