siRNA Details
Gene Symbol: FRMPD4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014728.3 | NP_055543.2 | 5 | 963 | ||
XM_005274632.3 | XP_005274689.2 | 5 | 641 | |||
XM_011545613.2 | XP_011543915.1 | 5 | 950 | |||
XM_017029983.1 | XP_016885472.1 | 5 | 853 | |||
XM_017029984.1 | XP_016885473.1 | 5 | 642 | |||
XM_017029985.1 | XP_016885474.1 | 5 | 639 | |||
XM_017029986.1 | XP_016885475.1 | 5 | 852 | |||
GenBank mRNA | AB002314.3 | - | 5 | 963 | ||
AK289694.1 | - | 5 | 950 | |||
BC113700.1 | - | 5 | 548 | 8322752 | ||
BC113702.1 | - | 5 | 548 | 8322754 | ||
BC143746.1 | - | 6 | 645 | 9052260 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
