siRNA Details
Gene Symbol: TCEAL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001006639.1 | NP_001006640.1 | 3 | 684 | ||
NM_001006640.1 | NP_001006641.1 | 3 | 696 | |||
NM_004780.2 | NP_004771.2 | 3 | 696 | |||
GenBank mRNA | BC000809.1 | - | 3 | 649 | 3452909 | |
BX092470.1 | - | 3 | 667 | |||
CF454849.1 | - | 3 | 672 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
