siRNA Details
Gene Symbol: HOMER2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004839.3 | NP_004830.2 | 9 | 1688 | ||
NM_199330.2 | NP_955362.1 | 9 | 1721 | |||
XM_005272448.4 | XP_005272505.1 | 9 | 2156 | |||
XM_005272449.3 | XP_005272506.1 | 9 | 1568 | |||
XM_006720776.3 | XP_006720839.1 | 9 | 1624 | |||
XM_011522231.2 | XP_011520533.1 | 9 | 2040 | |||
XM_011522233.2 | XP_011520535.1 | 9 | 1571 | |||
XM_011522234.2 | XP_011520536.1 | 9 | 1838 | |||
GenBank mRNA | AF093263.1 | - | 8 | 1499 | ||
AF093264.1 | - | 8 | 1532 | |||
BC012109.1 | - | 9 | 1685 | 4636423 | ||
Y19025.1 | - | 8 | 1499 | |||
Y19026.1 | - | 8 | 1532 | |||
Y19027.1 | - | 8 | 1335 | |||
Y19028.1 | - | 8 | 1368 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
