siRNA Details
Gene Symbol: KMT5B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001300907.1 | NP_001287836.1 | 2 | 341 | ||
NM_001300908.1 | NP_001287837.1 | 2 | 429 | |||
NM_001300909.1 | NP_001287838.1 | 2 | 429 | |||
NM_016028.4 | NP_057112.3 | 2 | 313 | |||
NM_017635.4 | NP_060105.3 | 2 | 341 | |||
XM_005274035.3 | XP_005274092.2 | 2 | 561 | |||
XM_005274036.3 | XP_005274093.2 | 2 | 561 | |||
XM_006718581.1 | XP_006718644.1 | 2 | 292 | |||
XM_011545091.1 | XP_011543393.1 | 1 | 135 | |||
XM_011545093.2 | XP_011543395.1 | 2 | 561 | |||
XM_011545094.2 | XP_011543396.1 | 2 | 561 | |||
XM_017017876.1 | XP_016873365.1 | 2 | 561 | |||
XM_017017877.1 | XP_016873366.1 | 2 | 561 | |||
XM_017017878.1 | XP_016873367.1 | 2 | 292 | |||
XM_017017879.1 | XP_016873368.1 | 2 | 307 | |||
XM_017017880.1 | XP_016873369.1 | 2 | 561 | |||
GenBank mRNA | AF151843.1 | - | 1 | 55 | ||
AF264782.1 | - | 1 | 233 | |||
AK299201.1 | - | 2 | 258 | |||
AK314336.1 | - | 2 | 256 | |||
BC002522.2 | - | 2 | 260 | |||
BC065287.1 | - | 2 | 265 | |||
BC087834.1 | - | 2 | 282 | |||
BC098121.1 | - | 1 | 62 | 40001705 | ||
BC099714.1 | - | 1 | 62 | 40001707 | ||
BC103498.1 | - | 1 | 62 | 40001706 | ||
BC104483.1 | - | 1 | 62 | 40001704 | ||
BM152996.1 | - | 2 | 314 | |||
CR997801.1 | - | 2 | 429 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
