siRNA Details
Gene Symbol: DSCAM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271534.1 | NP_001258463.1 | 4 | 1021 | ||
NM_001389.3 | NP_001380.2 | 4 | 1021 | |||
NR_073202.1 | 4 | 1021 | ||||
XM_017028281.1 | XP_016883770.1 | 1 | 76 | |||
GenBank mRNA | AB209136.1 | - | 4 | 604 | ||
AF023449.1 | - | 3 | 527 | |||
AF023450.1 | - | 4 | 1021 | |||
AF217525.1 | - | 4 | 1047 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Set Membership: |
![]() |
Gene Ontology Categories:
