siRNA Details
Gene Symbol: KNDC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152643.6 | NP_689856.6 | 26 | 4589 | ||
XM_017016858.1 | XP_016872347.1 | 26 | 4957 | |||
GenBank mRNA | AB000781.1 | - | 25 | 4485 | ||
AK131205.1 | - | 7 | 870 | |||
AK294170.1 | - | 9 | 1183 | |||
BQ719533.1 | - | 6 | 727 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
