siRNA Details
Gene Symbol: KNDC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152643.6 | NP_689856.6 | 6 | 1007 | ||
XM_017016858.1 | XP_016872347.1 | 6 | 1375 | |||
XM_017016859.1 | XP_016872348.1 | 6 | 1378 | |||
GenBank mRNA | AB000781.1 | - | 5 | 903 | ||
AB051555.1 | - | 5 | 1101 | |||
AK090457.1 | - | 6 | 970 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
