siRNA Details
Gene Symbol: MAP6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033063.1 | NP_149052.1 | 4 | 1488 | ||
XM_017017755.1 | XP_016873244.1 | 5 | 4075 | |||
XM_017017756.1 | XP_016873245.1 | 5 | 638 | |||
XM_017017757.1 | XP_016873246.1 | 5 | 657 | |||
XM_017017758.1 | XP_016873247.1 | 5 | 672 | |||
GenBank mRNA | AK123340.1 | - | 4 | 596 | ||
BC063860.1 | - | 4 | 1154 | |||
BC139780.1 | - | 5 | 727 | 40122938 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
