siRNA Details
Gene Symbol: MAP6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033063.1 | NP_149052.1 | 4 | 1751 | ||
XM_017017755.1 | XP_016873244.1 | 5 | 4338 | |||
XM_017017756.1 | XP_016873245.1 | 5 | 901 | |||
XM_017017757.1 | XP_016873246.1 | 5 | 920 | |||
XM_017017758.1 | XP_016873247.1 | 5 | 935 | |||
GenBank mRNA | AK123340.1 | - | 4 | 859 | ||
BC063860.1 | - | 4 | 1417 | |||
BC139780.1 | - | 5 | 990 | 40122938 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
