siRNA Details
Gene Symbol: ETV3L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004341.2 | NP_001004341.1 | 5 | 1618 | ||
GenBank mRNA | AK131392.1 | - | 5 | 1618 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
