siRNA Details
Gene Symbol: RAD52 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001297419.1 | NP_001284348.1 | 11, 12 | 1444 | ||
NM_001297421.1 | NP_001284350.1 | 9, 10 | 1128 | |||
NM_134424.3 | NP_602296.2 | 11, 12 | 1332 | |||
NR_123713.1 | 13, 14 | 1614 | ||||
XM_005253720.4 | XP_005253777.1 | 12, 13 | 2206 | |||
XM_005253721.2 | XP_005253778.1 | 11, 12 | 1432 | |||
XM_011520990.2 | XP_011519292.1 | 12, 13 | 1468 | |||
XM_011520991.2 | XP_011519293.1 | 11, 12 | 1334 | |||
XM_011520995.2 | XP_011519297.1 | 7, 8 | 916 | |||
XM_017019769.1 | XP_016875258.1 | 12, 13 | 1589 | |||
XM_017019770.1 | XP_016875259.1 | 10, 11 | 1181 | |||
GenBank mRNA | AF125948.1 | - | 10, 11 | 1504 | ||
AF125949.1 | - | 11, 12 | 1728 | |||
AK290047.1 | - | 11, 12 | 1252 | |||
AK299283.1 | - | 9, 10 | 1075 | |||
AY540753.1 | - | 10, 11 | 1194 | |||
BC144458.1 | - | 10, 11 | 1532 | |||
L33262.1 | - | 11, 12 | 1462 | |||
U12134.1 | - | 10, 11 | 1225 | |||
U27516.1 | - | 10, 11 | 1224 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
