siRNA Details
Gene Symbol: RAD52 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001297419.1 | NP_001284348.1 | 12 | 2129 | ||
NM_001297421.1 | NP_001284350.1 | 10 | 1813 | |||
NM_134424.3 | NP_602296.2 | 12 | 2017 | |||
NR_123713.1 | 14 | 2299 | ||||
XM_005253720.4 | XP_005253777.1 | 13 | 2891 | |||
XM_005253721.2 | XP_005253778.1 | 12 | 2117 | |||
XM_011520990.2 | XP_011519292.1 | 13 | 2153 | |||
XM_011520991.2 | XP_011519293.1 | 12 | 2019 | |||
XM_011520995.2 | XP_011519297.1 | 8 | 1601 | |||
XM_017019769.1 | XP_016875258.1 | 13 | 2274 | |||
XM_017019770.1 | XP_016875259.1 | 11 | 1866 | |||
GenBank mRNA | AF125948.1 | - | 11 | 2191 | ||
AF125949.1 | - | 12 | 2415 | |||
BC144458.1 | - | 11 | 2217 | |||
L33262.1 | - | 12 | 2147 | |||
U12134.1 | - | 11 | 1910 | |||
U27516.1 | - | 11 | 1909 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
