siRNA Details
Gene Symbol: RAD52 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001297419.1 | NP_001284348.1 | 7, 8 | 786 | ||
NM_001297421.1 | NP_001284350.1 | 5, 6 | 470 | |||
NM_001297422.1 | NP_001284351.1 | 7, 8 | 674 | |||
NM_134424.3 | NP_602296.2 | 7, 8 | 674 | |||
NR_123713.1 | 9, 10 | 956 | ||||
XM_005253720.4 | XP_005253777.1 | 8, 9 | 1548 | |||
XM_005253721.2 | XP_005253778.1 | 7, 8 | 774 | |||
XM_011520990.2 | XP_011519292.1 | 8, 9 | 810 | |||
XM_011520991.2 | XP_011519293.1 | 7, 8 | 676 | |||
XM_011520993.2 | XP_011519295.1 | 7, 8 | 625 | |||
XM_011520995.2 | XP_011519297.1 | 3, 4 | 258 | |||
XM_017019769.1 | XP_016875258.1 | 8, 9 | 931 | |||
XM_017019770.1 | XP_016875259.1 | 7, 8 | 625 | |||
XM_017019771.1 | XP_016875260.1 | 7, 8 | 625 | |||
GenBank mRNA | AF125948.1 | - | 6, 7 | 797 | ||
AF125949.1 | - | 7, 8 | 1050 | |||
AK290047.1 | - | 7, 8 | 594 | |||
AK294709.1 | - | 7, 8 | 619 | |||
AK299283.1 | - | 5, 6 | 417 | |||
AY540753.1 | - | 6, 7 | 536 | |||
BC104015.1 | - | 7, 8 | 617 | |||
BC104016.1 | - | 7, 8 | 695 | |||
BC104017.1 | - | 7, 8 | 617 | |||
BC114507.1 | - | 6, 7 | 581 | |||
BC114954.1 | - | 8, 9 | 820 | |||
BC144458.1 | - | 6, 7 | 874 | |||
L33262.1 | - | 7, 8 | 804 | |||
U12134.1 | - | 6, 7 | 567 | |||
U27516.1 | - | 6, 7 | 566 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
