siRNA Details
Gene Symbol: CHSY3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175856.4 | NP_787052.3 | 3 | 2258 | ||
XM_011543363.2 | XP_011541665.1 | 3 | 2111 | |||
XM_011543364.2 | XP_011541666.1 | 2 | 1467 | |||
XM_017009434.1 | XP_016864923.1 | 2 | 11157 | |||
GenBank mRNA | AB086062.1 | - | 3 | 2258 | ||
AB175496.1 | - | 3 | 2258 | |||
AJ504664.2 | - | 3 | 1795 | |||
AJ578034.2 | - | 3 | 2616 | |||
AY726604.1 | - | 3 | 1560 | |||
BC137325.1 | - | 3 | 2327 | 9021322 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
