siRNA Details
Gene Symbol: PGAM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001317079.1 | NP_001304008.1 | 4 | 1463 | ||
NM_002629.3 | NP_002620.1 | 4 | 1533 | |||
GenBank mRNA | AK223096.1 | - | 4 | 1379 | ||
AK292216.1 | - | 4 | 1434 | |||
AK295991.1 | - | 5 | 1269 | |||
AK297491.1 | - | 5 | 1279 | |||
AK315910.1 | - | 4 | 1466 | |||
AY007118.1 | - | 1 | 1370 | |||
BC000455.1 | - | 1 | 1396 | 2821567 | ||
BC010038.1 | - | 1 | 1388 | 3604026 | ||
BC011678.2 | - | 1 | 1352 | 4299008 | ||
BC053356.1 | - | 1 | 1369 | 5787678 | ||
BC062302.1 | - | 1 | 1368 | 6584466 | ||
BC066959.1 | - | 4 | 1401 | 5262935 | ||
BC073742.1 | - | 1 | 1367 | 6297173 | ||
BC157873.1 | - | 4 | 1385 | 8862699 | ||
J04173.1 | - | 4 | 1381 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
