siRNA Details
Gene Symbol: ITIH5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001851.2 | NP_001001851.1 | 10 | 1764 | ||
NM_030569.6 | NP_085046.5 | 10 | 1764 | |||
NM_032817.5 | NP_116206.4 | 6 | 1099 | |||
XM_011519713.2 | XP_011518015.1 | 11 | 1839 | |||
XM_011519714.2 | XP_011518016.1 | 11 | 1839 | |||
GenBank mRNA | AB075833.1 | - | 7 | 1305 | ||
AF318347.1 | - | 2 | 649 | |||
AK027375.1 | - | 10 | 1701 | |||
AK027831.1 | - | 7 | 1283 | |||
AK027849.1 | - | 5 | 948 | |||
AK075381.1 | - | 10 | 1737 | |||
AL833203.1 | - | 10 | 1609 | |||
AY238437.1 | - | 10 | 1660 | |||
AY358426.1 | - | 10 | 1729 | |||
BC136439.1 | - | 10 | 1737 | |||
CR627109.1 | - | 5 | 6198 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
