siRNA Details
Gene Symbol: FGF13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001139498.1 | NP_001132970.1 | 6 | 1462 | ||
NM_001139500.1 | NP_001132972.1 | 7 | 1630 | |||
NM_001139501.1 | NP_001132973.1 | 8 | 1740 | |||
NM_001139502.1 | NP_001132974.1 | 6 | 1383 | |||
NM_004114.3 | NP_004105.1 | 5 | 1995 | |||
NM_033642.2 | NP_378668.1 | 5 | 1258 | |||
XM_005262399.1 | XP_005262456.1 | 7 | 1833 | |||
GenBank mRNA | AF100143.1 | - | 5 | 1564 | ||
AF100144.1 | - | 5 | 1259 | |||
BC012347.1 | - | 5 | 1992 | 4551396 | ||
BC034340.1 | - | 5 | 1699 | 3918908 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
