siRNA Details
Gene Symbol: FGF13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001139498.1 | NP_001132970.1 | 6 | 862 | ||
NM_001139500.1 | NP_001132972.1 | 7 | 1030 | |||
NM_001139501.1 | NP_001132973.1 | 8 | 1140 | |||
NM_001139502.1 | NP_001132974.1 | 6 | 783 | |||
NM_004114.3 | NP_004105.1 | 5 | 1395 | |||
NM_033642.2 | NP_378668.1 | 5 | 658 | |||
XM_005262399.1 | XP_005262456.1 | 7 | 1233 | |||
GenBank mRNA | AF100143.1 | - | 5 | 963 | ||
AF100144.1 | - | 5 | 658 | |||
AK289960.1 | - | 5 | 1073 | |||
AK297545.1 | - | 7 | 1061 | |||
AK303685.1 | - | 6 | 862 | |||
AK316170.1 | - | 6 | 939 | |||
BC012347.1 | - | 5 | 1392 | 4551396 | ||
BC034340.1 | - | 5 | 1099 | 3918908 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
