siRNA Details
Gene Symbol: KIAA1024 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015206.2 | NP_056021.1 | 4 | 5638 | ||
XM_011521392.1 | XP_011519694.1 | 4 | 5677 | |||
XM_011521393.2 | XP_011519695.1 | 4 | 8513 | |||
XM_011521395.2 | XP_011519697.1 | 4 | 6123 | |||
XM_011521396.2 | XP_011519698.1 | 4 | 5848 | |||
XM_017022027.1 | XP_016877516.1 | 4 | 5697 | |||
GenBank mRNA | AB028947.2 | - | 4 | 5638 | ||
AF035294.1 | - | 1 | 995 | |||
BC152466.1 | - | 4 | 5634 | 8862602 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
