siRNA Details
Gene Symbol: TBL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012453.2 | NP_036585.1 | 1 | 207 | ||
XM_011516027.2 | XP_011514329.1 | 1 | 100 | |||
GenBank mRNA | AF056183.1 | - | 1 | 35 | ||
AF097484.1 | - | 1 | 92 | |||
AK000536.1 | - | 1 | 100 | |||
AK074619.1 | - | 1 | 103 | |||
AK130791.1 | - | 1 | 100 | |||
AK302277.1 | - | 1 | 123 | |||
AK314499.1 | - | 1 | 100 | |||
AL080162.1 | - | 1 | 167 | |||
AY358518.1 | - | 1 | 119 | |||
BC012938.1 | - | 1 | 84 | 4453977 | ||
BC047354.1 | - | 1 | 145 | |||
BX420755.2 | - | 1 | 208 | |||
KU178597.1 | - | 1 | 73 | |||
KU178598.1 | - | 1 | 73 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
